Assay Details
Target Gene Details
Entrez Gene ID: | 60526 |
Gene Name: | lipid droplet associated hydrolase |
Gene Aliases: |
C2orf43, hLDAH |
Location: |
Chr.2:20682489-20823139 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LDAH | NM_001282719.1 | NP_001269648.1 | ||
| NM_001282720.1 | NP_001269649.1 | |||
| NM_001282721.1 | NP_001269650.1 | |||
| NM_001282722.1 | NP_001269651.1 | |||
| NM_001282723.1 | NP_001269652.1 | |||
| NM_001282724.1 | NP_001269653.1 | |||
| NM_021925.3 | NP_068744.1 | |||
| NR_104233.1 | ||||
| XM_011533014.2 | XP_011531316.1 | |||
| AK021672.1 | ||||
| AK025473.1 | BAB15142.1 | |||
| AK299244.1 | ||||
| AK299555.1 | ||||
| AK300494.1 | ||||
| AK301493.1 | ||||
| AK316162.1 | ||||
| AK316204.1 | ||||
| AK316310.1 | ||||
| BC017473.1 | AAH17473.1 |
More Information
Set Membership: |
|

Genomic Map