Assay Details
Target Gene Details
Entrez Gene ID: | 79582 |
Gene Name: | sperm associated antigen 16 |
Gene Aliases: |
PF20, WDR29 |
Location: |
Chr.2:213284369-214410501 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3152 | Chr.2:213282926 - 213332692 on Build GRCh38 | Insertion |
|
| nsv1013206 | Chr.2:213267855 - 213302172 on Build GRCh38 | Loss |
|
| nsv834531 | Chr.2:213250900 - 213437519 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map