Assay Details
Target Gene Details
Entrez Gene ID: | 5270 |
Gene Name: | serpin family E member 2 |
Gene Aliases: |
GDN, GDNPF, PI-7, PI7, PN-1, PN1, PNI |
Location: |
Chr.2:223975048-224039319 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SERPINE2 | NM_001136528.1 | NP_001130000.1 | ||
NM_001136530.1 | NP_001130002.1 | |||
NM_006216.3 | NP_006207.1 | |||
NR_073116.1 | ||||
XM_005246641.2 | XP_005246698.1 | |||
XM_017004329.1 | XP_016859818.1 | |||
XM_017004330.1 | XP_016859819.1 | |||
XM_017004331.1 | XP_016859820.1 | |||
XM_017004332.1 | XP_016859821.1 | |||
A03911.1 | CAA00310.1 | |||
AK225823.1 | ||||
AK295564.1 | ||||
AK297588.1 | ||||
AK312499.1 | ||||
BC015663.2 | AAH15663.1 | |||
BC042628.1 | AAH42628.1 | |||
M17783.1 | AAA35883.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
