Assay Details
Target Gene Details
Entrez Gene ID: | 26010 |
Gene Name: | spermatogenesis associated serine rich 2 like |
Gene Aliases: |
DNAPTP6, SGNP |
Location: |
Chr.2:200305881-200482263 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
