Assay Details
Target Gene Details
Entrez Gene ID: | 9581 |
Gene Name: | prolyl endopeptidase-like |
Gene Aliases: |
- |
Location: |
Chr.2:44317607-44361862 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PREPL | NM_001042385.2 | NP_001035844.1 | ||
NM_001042386.2 | NP_001035845.1 | |||
NM_001171603.1 | NP_001165074.1 | |||
NM_001171606.1 | NP_001165077.1 | |||
NM_001171613.1 | NP_001165084.1 | |||
NM_001171617.1 | NP_001165088.1 | |||
NM_006036.4 | NP_006027.2 | |||
XM_011533198.1 | XP_011531500.1 | |||
XM_011533200.1 | XP_011531502.1 | |||
XM_011533202.1 | XP_011531504.1 | |||
XM_017005384.1 | XP_016860873.1 | |||
XM_017005385.1 | XP_016860874.1 | |||
AB007896.1 | ||||
AK025649.1 | ||||
AK131463.1 | BAD18608.1 | |||
BC151236.1 | ||||
DQ023503.1 | AAY89634.1 | |||
DQ023504.1 | AAY89635.1 | |||
DQ023505.1 | AAY89636.1 | |||
DQ023506.1 | AAY89637.1 | |||
DQ023507.1 | AAY89638.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv3798n100 | Chr.2:44279994 - 44404126 on Build GRCh38 | Loss |
![]() ![]() ![]() |
dgv6837n54 | Chr.2:44278046 - 44351759 on Build GRCh38 | Loss |
![]() ![]() |
dgv6838n54 | Chr.2:44297436 - 44341555 on Build GRCh38 | Loss |
![]() ![]() |
nsv1006405 | Chr.2:44013164 - 44709137 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
esv3892026 | Chr.2:44318437 - 44418772 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv1011655 | Chr.2:44278899 - 44344334 on Build GRCh38 | Loss |
![]() ![]() |
nsv457196 | Chr.2:44276990 - 44339733 on Build GRCh38 | Gain |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
