Assay Details
Target Gene Details
Entrez Gene ID: | 80255 |
Gene Name: | solute carrier family 35 member F5 |
Gene Aliases: |
- |
Location: |
Chr.2:113702677-113756823 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC35F5 | XM_017005027.1 | 17 | 3878 | XP_016860516.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3592016 | Chr.2:113233328 - 113804636 on Build GRCh38 | Gain |
|
| nsv1001195 | Chr.2:113604426 - 113748753 on Build GRCh38 | Gain |
|
| nsv2882 | Chr.2:113695137 - 113729107 on Build GRCh38 | Insertion |
|
| esv3592029 | Chr.2:113671263 - 113708339 on Build GRCh38 | Gain |
|
| esv3592028 | Chr.2:113650431 - 113744360 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map