Assay Details
Target Gene Details
Entrez Gene ID: | 90 |
Gene Name: | activin A receptor type 1 |
Gene Aliases: |
ACTRI, ACVR1A, ACVRLK2, ALK2, FOP, SKR1, TSRI |
Location: |
Chr.2:157736446-157875880 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACVR1 | NM_001105.4 | NP_001096.1 | ||
| NM_001111067.2 | NP_001104537.1 | |||
| XM_005246939.3 | XP_005246996.1 | |||
| XM_005246940.3 | XP_005246997.1 | |||
| XM_006712825.3 | XP_006712888.1 | |||
| XM_011512106.2 | XP_011510408.1 | |||
| XM_011512107.2 | XP_011510409.1 | |||
| XM_011512108.2 | XP_011510410.1 | |||
| AK313630.1 | ||||
| BC033867.1 | AAH33867.1 | |||
| KU177885.1 | ||||
| KU177886.1 | ||||
| KU177887.1 | ||||
| L02911.1 | AAA36614.1 | |||
| Z22534.1 | CAA80256.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114192401] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map