Assay Details
Target Gene Details
Entrez Gene ID: | 64969 |
Gene Name: | mitochondrial ribosomal protein S5 |
Gene Aliases: |
MRP-S5, S5mt |
Location: |
Chr.2:95085391-95122027 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MRPS5 | NM_001321995.1 | NP_001308924.1 | ||
NM_001321996.1 | NP_001308925.1 | |||
NM_001321997.1 | NP_001308926.1 | |||
NM_031902.4 | NP_114108.1 | |||
NR_136152.1 | ||||
NR_136153.1 | ||||
AB049940.1 | BAB40993.1 | |||
AK058160.1 | BAB71695.1 | |||
AK295812.1 | ||||
BC014172.2 | AAH14172.1 | |||
BG771419.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1160825 | Chr.2:95050220 - 95445574 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3591662 | Chr.2:94986856 - 95146757 on Build GRCh38 | Gain |
![]() ![]() ![]() |
dgv4007n100 | Chr.2:94662150 - 95248719 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
