Assay Details
Target Gene Details
Entrez Gene ID: | 56896 |
Gene Name: | dihydropyrimidinase like 5 |
Gene Aliases: |
CRAM, CRMP-5, CRMP5, Ulip6 |
Location: |
Chr.2:26848101-26950351 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPYSL5 | NM_001253723.1 | NP_001240652.1 | ||
| NM_001253724.1 | NP_001240653.1 | |||
| NM_020134.3 | NP_064519.2 | |||
| AF264015.1 | AAK16830.1 | |||
| BC002874.2 | AAH02874.1 | |||
| DA410946.1 |
More Information
Set Membership: |
|

Genomic Map