Assay Details
Target Gene Details
Entrez Gene ID: | 8291 |
Gene Name: | dysferlin |
Gene Aliases: |
FER1L1, LGMD2B, MMD1 |
Location: |
Chr.2:71453155-71686763 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DYSF | NM_001130976.1 | NP_001124448.1 | ||
NM_001130977.1 | NP_001124449.1 | |||
NM_001130978.1 | NP_001124450.1 | |||
NM_001130979.1 | NP_001124451.1 | |||
NM_001130980.1 | NP_001124452.1 | |||
NM_001130981.1 | NP_001124453.1 | |||
NM_003494.3 | NP_003485.1 | |||
XM_005264585.4 | XP_005264642.1 | |||
AF075575.1 | AAC63519.1 | |||
EU515155.1 | ||||
EU515156.1 | ||||
EU515157.1 | ||||
EU515158.1 | ||||
EU515159.1 | ||||
EU515160.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
