Assay Details
Target Gene Details
Entrez Gene ID: | 129804 |
Gene Name: | fibulin 7 |
Gene Aliases: |
TM14 |
Location: |
Chr.2:112138385-112254888 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBLN7 |
Target Gene Details
Entrez Gene ID: | 84524 |
Gene Name: | zinc finger CCCH-type containing 8 |
Gene Aliases: |
Fliz1, ZC3HDC8 |
Location: |
Chr.2:112215035-112255087 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZC3H8 | NM_032494.2 | NP_115883.2 | ||
| XM_011511998.1 | XP_011510300.1 | |||
| XM_017005107.1 | XP_016860596.1 | |||
| AF334161.1 | AAK13496.1 | |||
| AK315329.1 | ||||
| BC032001.1 | AAH32001.1 | |||
| BG773518.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521956 | Chr.2:110634682 - 112341235 on Build GRCh38 | Loss |
|
| nsv582703 | Chr.2:112162642 - 112342437 on Build GRCh38 | Loss |
|
| dgv4047n100 | Chr.2:110308696 - 112358403 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map