Assay Details
Target Gene Details
Entrez Gene ID: | 116255 |
Gene Name: | monoacylglycerol O-acyltransferase 1 |
Gene Aliases: |
DGAT2L, DGAT2L1, MGAT1 |
Location: |
Chr.2:222671738-222709930 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOGAT1 | NM_058165.2 | NP_477513.2 | ||
| AF384163.1 | AAK84178.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3594427 | Chr.2:222666972 - 222675602 on Build GRCh38 | Loss |
|
| nsv1011830 | Chr.2:222673597 - 222979600 on Build GRCh38 | Gain |
|
| nsv834551 | Chr.2:222607248 - 222776534 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map