Assay Details
Target Gene Details
Entrez Gene ID: | 9541 |
Gene Name: | corepressor interacting with RBPJ, 1 |
Gene Aliases: |
CIR |
Location: |
Chr.2:174348150-174395715 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CIR1 | NM_004882.3 | NP_004873.3 | ||
AF098297.1 | AAD05243.1 | |||
AK292389.1 | ||||
BC021175.1 | AAH21175.1 | |||
BC031018.1 | ||||
BC038987.1 | AAH38987.1 | |||
BC046098.2 | AAH46098.1 | |||
BU596189.1 | ||||
U03644.1 | AAA17853.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
