Assay Details
Target Gene Details
Entrez Gene ID: | 79745 |
Gene Name: | CAP-Gly domain containing linker protein family member 4 |
Gene Aliases: |
RSNL2 |
Location: |
Chr.2:29097676-29183813 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLIP4 | NM_001287528.1 | NP_001274457.1 | ||
| XM_005264562.2 | XP_005264619.1 | |||
| XM_011533110.1 | XP_011531412.1 | |||
| AF433661.1 | AAP97312.1 | |||
| AK302325.1 | ||||
| DC317668.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34904 | Chr.2:28979483 - 30284483 on Build GRCh38 | Gain |
|
| nsv1008490 | Chr.2:28884768 - 29482018 on Build GRCh38 | Gain |
|
| esv2759035 | Chr.2:28953630 - 30316563 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map