Assay Details
Target Gene Details
Entrez Gene ID: | 26275 |
Gene Name: | 3-hydroxyisobutyryl-CoA hydrolase |
Gene Aliases: |
HIBYLCOAH |
Location: |
Chr.2:190204634-190320045 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIBCH | NM_014362.3 | NP_055177.2 | ||
| NM_198047.2 | NP_932164.1 | |||
| XM_011510953.1 | XP_011509255.1 | |||
| XM_011510954.1 | XP_011509256.1 | |||
| AI767349.1 | ||||
| AK126890.1 | ||||
| AK222979.1 | BAD96699.1 | |||
| AK223023.1 | BAD96743.1 | |||
| AK308042.1 | ||||
| AK313651.1 | ||||
| BC005190.1 | AAH05190.2 | |||
| BC067822.1 | AAH67822.1 | |||
| CA448468.1 | ||||
| U66669.1 | AAC52114.1 |
More Information
Set Membership: |
|

Genomic Map