Assay Details
Target Gene Details
Entrez Gene ID: | 80067 |
Gene Name: | DDB1 and CUL4 associated factor 17 |
Gene Aliases: |
C2orf37 |
Location: |
Chr.2:171434166-171491029 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF17 | NM_001164821.1 | NP_001158293.1 | ||
| NM_025000.3 | NP_079276.2 | |||
| NR_028482.1 | ||||
| XM_006712767.1 | XP_006712830.1 | |||
| XM_006712768.1 | XP_006712831.1 | |||
| XM_011511881.1 | XP_011510183.1 | |||
| XM_011511882.1 | XP_011510184.1 | |||
| XM_017004995.1 | XP_016860484.1 | |||
| XM_017004996.1 | XP_016860485.1 | |||
| XM_017004997.1 | XP_016860486.1 | |||
| XM_017004998.1 | XP_016860487.1 | |||
| XM_017004999.1 | XP_016860488.1 | |||
| XM_017005000.1 | XP_016860489.1 | |||
| XM_017005001.1 | XP_016860490.1 | |||
| XM_017005002.1 | XP_016860491.1 | |||
| AK023158.1 | BAB14436.1 | |||
| AK307877.1 | ||||
| BC140843.1 | ||||
| CR933646.1 | CAI45947.1 |
More Information
Set Membership: |
|

Genomic Map