Assay Details
Target Gene Details
Entrez Gene ID: | 57142 |
Gene Name: | reticulon 4 |
Gene Aliases: |
ASY, NI220/250, NOGO, NOGO-A, NOGOC, NSP, NSP-CL, Nbla00271, Nbla10545, Nogo-B, Nogo-C, RTN-X, RTN4-A, RTN4-B1, RTN4-B2, RTN4-C |
Location: |
Chr.2:54972187-55137831 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2720078 | Chr.2:53962225 - 55281329 on Build GRCh38 | Deletion |
|
| nsv834069 | Chr.2:54869460 - 55024544 on Build GRCh38 | Loss |
|
| nsv953127 | Chr.2:54970765 - 55049264 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map