Assay Details
Target Gene Details
Entrez Gene ID: | 80895 |
Gene Name: | ILK associated serine/threonine phosphatase |
Gene Aliases: |
ILKAP2, ILKAP3, PP2C-DELTA, PPM1O |
Location: |
Chr.2:238170401-238203702 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ILKAP | NM_030768.2 | NP_110395.1 | ||
| XM_005246106.1 | XP_005246163.1 | |||
| XM_006712784.1 | XP_006712847.1 | |||
| XM_011511946.2 | XP_011510248.1 | |||
| XM_017005056.1 | XP_016860545.1 | |||
| XM_017005057.1 | XP_016860546.1 | |||
| XM_017005058.1 | XP_016860547.1 | |||
| AK001043.1 | ||||
| AK055417.1 | ||||
| AL136850.1 | CAB66784.1 | |||
| AY024365.1 | AAK07736.1 | |||
| BC006576.2 | AAH06576.1 | |||
| CR533533.1 | CAG38564.1 | |||
| FJ491380.1 | ||||
| HQ658059.1 | ||||
| HQ658060.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map