Assay Details
Target Gene Details
Entrez Gene ID: | 56171 |
Gene Name: | dynein axonemal heavy chain 7 |
Gene Aliases: |
- |
Location: |
Chr.2:195737604-196075188 on Build GRCh38 |
Assay Gene Location: | Within Intron 61 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DNAH7 | NM_018897.2 | NP_061720.2 | ||
XM_011511487.2 | XP_011509789.1 | |||
XM_011511488.2 | XP_011509790.1 | |||
XM_011511489.2 | XP_011509791.1 | |||
XM_011511490.2 | XP_011509792.1 | |||
XM_011511497.2 | XP_011509799.1 | |||
XM_017004504.1 | XP_016859993.1 | |||
AB023161.2 | ||||
AF327442.1 | AAL37427.1 | |||
BC029567.1 | AAH29567.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -


Gene Ontology Categories:
