Assay Details
Target Gene Details
Entrez Gene ID: | 1876 |
Gene Name: | E2F transcription factor 6 |
Gene Aliases: |
E2F-6 |
Location: |
Chr.2:11444375-11466177 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E2F6 | NM_001278275.1 | NP_001265204.1 | ||
| NM_001278276.1 | NP_001265205.1 | |||
| NM_001278277.1 | NP_001265206.1 | |||
| NM_001278278.1 | NP_001265207.1 | |||
| NM_198256.3 | NP_937987.2 | |||
| NM_212540.2 | NP_997705.1 | |||
| NR_103490.1 | ||||
| XM_017003547.1 | 6 | 1393 | XP_016859036.1 | |
| XM_017003548.1 | 7 | 1480 | XP_016859037.1 | |
| XM_017003549.1 | 5 | 1338 | XP_016859038.1 | |
| AF041381.1 | AAC14694.1 | |||
| AF059292.1 | AAC31426.1 | |||
| AK096197.1 | ||||
| AK223159.1 | BAD96879.1 | |||
| AK223407.1 | BAD97127.1 | |||
| AK290413.1 | ||||
| AK293504.1 | ||||
| AY083996.1 | AAM10783.1 | |||
| AY551346.1 | AAT02636.1 | |||
| AY551347.1 | AAT02637.1 | |||
| AY551348.1 | AAT02638.1 | |||
| AY551349.1 | AAT02639.1 | |||
| AY551350.1 | AAT02640.1 | |||
| AY551351.1 | AAT02641.1 | |||
| BC008348.1 | AAH08348.1 | |||
| BC107740.1 | AAI07741.1 | |||
| BI772069.1 | ||||
| KU178035.1 | ||||
| KU178036.1 |
More Information
Set Membership: |
|

Genomic Map