Assay Details
Target Gene Details
Entrez Gene ID: | 9874 |
Gene Name: | tousled like kinase 1 |
Gene Aliases: |
PKU-beta |
Location: |
Chr.2:170990823-171231314 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLK1 | NM_001136554.1 | NP_001130026.1 | ||
| NM_001136555.1 | NP_001130027.1 | |||
| NM_012290.4 | NP_036422.3 | |||
| DN604386.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3593343 | Chr.2:170964107 - 170991716 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map