Assay Details
Target Gene Details
Entrez Gene ID: | 101929680 |
Gene Name: | uncharacterized LOC101929680 |
Gene Aliases: |
- |
Location: |
Chr.2:166081531-166301784 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929680 | NR_110260.1 | |||
| KM096550.1 | ||||
| KM096551.1 |
Target Gene Details
Entrez Gene ID: | 6323 |
Gene Name: | sodium voltage-gated channel alpha subunit 1 |
Gene Aliases: |
EIEE6, FEB3, FEB3A, FHM3, GEFSP2, HBSCI, NAC1, Nav1.1, SCN1, SMEI |
Location: |
Chr.2:165989160-166149216 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCN1A | NM_001202435.1 | NP_001189364.1 | ||
| XM_011511602.2 | XP_011509904.1 | |||
| XM_011511605.2 | XP_011509907.1 | |||
| XM_011511606.2 | XP_011509908.1 | |||
| XM_017004644.1 | XP_016860133.1 | |||
| XM_017004645.1 | XP_016860134.1 | |||
| XM_017004646.1 | XP_016860135.1 | |||
| XM_017004647.1 | XP_016860136.1 | |||
| XM_017004648.1 | XP_016860137.1 | |||
| XM_017004649.1 | XP_016860138.1 | |||
| XM_017004650.1 | XP_016860139.1 | |||
| XM_017004651.1 | XP_016860140.1 | |||
| XM_017004652.1 | XP_016860141.1 | |||
| XM_017004653.1 | XP_016860142.1 | |||
| XM_017004654.1 | XP_016860143.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114688154] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map