Assay Details
Target Gene Details
Entrez Gene ID: | 285175 |
Gene Name: | unc-80 homolog, NALCN activator |
Gene Aliases: |
C2orf21, UNC-80 |
Location: |
Chr.2:209771925-209999300 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
More Information
Additional Information:
For this assay, SNP(s) [rs73074801,rs73076704] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map