Assay Details
Target Gene Details
Entrez Gene ID: | 3267 |
Gene Name: | ArfGAP with FG repeats 1 |
Gene Aliases: |
HRB, RAB, RIP |
Location: |
Chr.2:227472169-227561222 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGFG1 | NM_001135187.1 | NP_001128659.1 | ||
| NM_001135188.1 | NP_001128660.1 | |||
| NM_001135189.1 | NP_001128661.1 | |||
| NM_004504.4 | NP_004495.2 | |||
| XM_005246516.3 | XP_005246573.1 | |||
| XM_006712478.3 | XP_006712541.1 | |||
| XM_006712479.3 | XP_006712542.1 | |||
| XM_006712480.3 | XP_006712543.1 | |||
| AK097451.1 | ||||
| BC030592.2 | AAH30592.1 | |||
| BC096272.3 | AAH96272.1 | |||
| BC096273.3 | AAH96273.1 | |||
| BC096274.1 | AAH96274.1 | |||
| BC096275.3 | AAH96275.1 | |||
| DB456989.1 | ||||
| KU178123.1 | ||||
| KU178124.1 | ||||
| KU178125.1 | ||||
| KU178126.1 | ||||
| X89478.1 | CAA61667.1 |
More Information
Set Membership: |
|

Genomic Map