Assay Details
Target Gene Details
Entrez Gene ID: | 130749 |
Gene Name: | carboxypeptidase O |
Gene Aliases: |
- |
Location: |
Chr.2:206932853-206971064 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPO | XM_017003372.1 | 2 | 4061 | XP_016858861.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011347 | Chr.2:206384007 - 207085773 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map