Assay Details
Target Gene Details
Entrez Gene ID: | 376940 |
Gene Name: | zinc finger CCCH-type containing 6 |
Gene Aliases: |
ZC3HDC6 |
Location: |
Chr.2:112275594-112340063 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZC3H6 | NM_198581.2 | NP_940983.2 | ||
| XM_006712519.2 | XP_006712582.1 | |||
| AB111887.1 | BAC98375.1 | |||
| AK131416.1 | BAD18563.1 | |||
| BC155540.1 | ||||
| BC167154.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521956 | Chr.2:110634682 - 112341235 on Build GRCh38 | Loss |
|
| nsv582703 | Chr.2:112162642 - 112342437 on Build GRCh38 | Loss |
|
| dgv4047n100 | Chr.2:110308696 - 112358403 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map