Assay Details
Target Gene Details
Entrez Gene ID: | 117583 |
Gene Name: | par-3 family cell polarity regulator beta |
Gene Aliases: |
ALS2CR19, PAR3B, PAR3L, PAR3beta |
Location: |
Chr.2:204545793-205620162 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARD3B | NM_001302769.1 | NP_001289698.1 | ||
| NM_057177.6 | NP_476518.4 | |||
| NM_152526.5 | NP_689739.4 | |||
| NM_205863.3 | NP_995585.2 | |||
| XM_017003291.1 | XP_016858780.1 | |||
| AB053321.1 | BAB69028.1 | |||
| AB073472.1 | BAC54035.1 | |||
| AB092439.1 | BAC54285.1 | |||
| AF428250.1 | AAL30664.1 | |||
| AF428251.1 | AAL30665.1 | |||
| AF466152.1 | AAL79827.1 | |||
| AI742400.1 | ||||
| AK056157.1 | BAB71106.1 | |||
| AW274720.1 | ||||
| BF434006.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893533 | Chr.2:204276082 - 204570913 on Build GRCh38 | Gain |
|
| nsv1001799 | Chr.2:202908732 - 205615330 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map