Assay Details
Target Gene Details
Entrez Gene ID: | 100507475 |
Gene Name: | IDH1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:208255233-208256194 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IDH1-AS1 | NR_046452.1 | 2 | 391 | |
| AI783767.1 | 1 | 103 | ||
| BJ994720.1 | 2 | 392 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3594112 | Chr.2:208255365 - 208261616 on Build GRCh38 | Loss |
|
| nsv584297 | Chr.2:208155457 - 208452035 on Build GRCh38 | Gain |
|
| nsv834517 | Chr.2:208142073 - 208305545 on Build GRCh38 | Loss |
|

Genomic Map