Assay Details
Target Gene Details
Entrez Gene ID: | 5937 |
Gene Name: | RNA binding motif single stranded interacting protein 1 |
Gene Aliases: |
C2orf12, HCC-4, MSSP, MSSP-1, MSSP-2, MSSP-3, SCR2, YC1 |
Location: |
Chr.2:160272151-160493856 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RBMS1 | NM_002897.4 | NP_002888.1 | ||
NM_016836.3 | NP_058520.1 | |||
XM_005246737.1 | XP_005246794.1 | |||
XM_005246738.2 | XP_005246795.1 | |||
XM_005246739.2 | XP_005246796.1 | |||
XM_005246740.1 | XP_005246797.1 | |||
XM_005246741.3 | XP_005246798.1 | |||
XM_017004626.1 | XP_016860115.1 | |||
AK127157.1 | ||||
AK225887.1 | ||||
AK297812.1 | ||||
BC012992.2 | AAH12992.1 | |||
BC012993.2 | AAH12993.1 | |||
BC018951.2 | AAH18951.1 | |||
BX648246.1 | ||||
D28482.1 | BAA05841.1 | |||
L11289.1 | ||||
X64652.1 | CAA45923.1 | |||
X77494.1 | CAA54628.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
