Assay Details
Target Gene Details
Entrez Gene ID: | 10267 |
Gene Name: | receptor activity modifying protein 1 |
Gene Aliases: |
- |
Location: |
Chr.2:237858714-237912117 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAMP1 | NM_001308353.1 | NP_001295282.1 | ||
| NM_005855.3 | NP_005846.1 | |||
| XM_017003152.1 | XP_016858641.1 | |||
| XM_017003153.1 | XP_016858642.1 | |||
| XM_017003155.1 | XP_016858644.1 | |||
| XM_017003156.1 | XP_016858645.1 | |||
| AI017108.1 | ||||
| AJ001014.1 | CAA04472.1 | |||
| AY265457.1 | AAP23298.1 | |||
| BC000548.2 | AAH00548.1 | |||
| BI548986.1 | ||||
| CR542032.1 | CAG46829.1 | |||
| CR542044.1 | CAG46841.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584754 | Chr.2:237865062 - 237894836 on Build GRCh38 | Loss |
|
| nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map