Assay Details
Target Gene Details
Entrez Gene ID: | 5937 |
Gene Name: | RNA binding motif single stranded interacting protein 1 |
Gene Aliases: |
C2orf12, HCC-4, MSSP, MSSP-1, MSSP-2, MSSP-3, SCR2, YC1 |
Location: |
Chr.2:160272151-160493856 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RBMS1 | NM_002897.4 | 14 | 2988 | NP_002888.1 |
NM_016836.3 | 14 | 2997 | NP_058520.1 | |
XM_005246737.1 | 15 | 2756 | XP_005246794.1 | |
XM_005246738.2 | 15 | 3091 | XP_005246795.1 | |
XM_005246739.2 | 15 | 2961 | XP_005246796.1 | |
XM_005246740.1 | 15 | 2747 | XP_005246797.1 | |
XM_005246741.3 | 15 | 2958 | XP_005246798.1 | |
XM_017004626.1 | 14 | 3034 | XP_016860115.1 | |
AI675628.1 | ||||
AL117617.1 | ||||
BC065192.1 | 1 | 258 | ||
BC069687.1 | 1 | 205 | AAH69687.1 | |
BC069706.1 | 1 | 204 | AAH69706.1 | |
BC069763.1 | 1 | 204 | AAH69763.1 | |
BC080620.1 | 1 | 232 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
