Assay Details
Target Gene Details
Entrez Gene ID: | 4436 |
Gene Name: | mutS homolog 2 |
Gene Aliases: |
COCA1, FCC1, HNPCC, HNPCC1, LCFS2 |
Location: |
Chr.2:47403067-47634501 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MSH2 | NM_000251.2 | NP_000242.1 | ||
| NM_001258281.1 | NP_001245210.1 | |||
| XM_005264332.3 | XP_005264389.2 | |||
| XM_011532867.1 | XP_011531169.1 | |||
| AK222860.1 | BAD96580.1 | |||
| AK223284.1 | BAD97004.1 | |||
| AK296831.1 | ||||
| AK297763.1 | ||||
| AK304496.1 | ||||
| AK311330.1 | ||||
| BC021566.1 | AAH21566.1 | |||
| BX649122.1 | ||||
| DC342099.1 | ||||
| DQ648894.1 | ||||
| L47574.1 | AAB59566.1 | |||
| L47575.1 | AAB59567.1 | |||
| L47576.1 | AAB59568.1 | |||
| L47577.1 | AAB59569.1 | |||
| L47578.1 | AAB59570.1 | |||
| L47579.1 | AAB59571.1 | |||
| L47580.1 | AAB59572.1 | |||
| L47581.1 | AAA76858.1 | |||
| L47582.1 | AAB59565.1 | |||
| L47583.1 | AAB59564.1 | |||
| U03911.1 | AAA18643.1 | |||
| U04045.1 | AAA61870.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv999576 | Chr.2:47278977 - 47546821 on Build GRCh38 | Gain |
|
| nsv1004828 | Chr.2:46829710 - 47587343 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map