Assay Details
Target Gene Details
Entrez Gene ID: | 6578 |
Gene Name: | solute carrier organic anion transporter family member 2A1 |
Gene Aliases: |
MATR1, OATP2A1, PGT, PHOAR2, SLC21A2 |
Location: |
Chr.3:133932696-134030076 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLCO2A1 | NM_005630.2 | NP_005621.2 | ||
| XM_017007075.1 | XP_016862564.1 | |||
| XM_017007076.1 | XP_016862565.1 | |||
| XM_017007077.1 | XP_016862566.1 | |||
| AK303550.1 | ||||
| BC041140.1 | AAH41140.2 | |||
| BC051347.1 | AAH51347.1 | |||
| U70867.1 | AAC09469.1 |
More Information
Set Membership: |
|

Genomic Map