Assay Details
Target Gene Details
Entrez Gene ID: | 26059 |
Gene Name: | ELKS/RAB6-interacting/CAST family member 2 |
Gene Aliases: |
CAST, CAST1, ELKSL, SPBC110, Spc110 |
Location: |
Chr.3:55508311-56468473 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
Target Gene Details
Entrez Gene ID: | 711 |
Gene Name: | ERC2 intronic transcript 1 |
Gene Aliases: |
C1orf1, C3orf51, Po42 |
Location: |
Chr.3:55657206-55659469 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERC2-IT1 | NR_024615.1 | 1 | 1517 | |
| U88965.1 | 1 | 1430 | AAC39932.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517851 | Chr.3:54375499 - 55705332 on Build GRCh38 | Gain |
|
| nsv1013405 | Chr.3:55040452 - 56437132 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map