Assay Details
Target Gene Details
Entrez Gene ID: | 344758 |
Gene Name: | G protein-coupled receptor 149 |
Gene Aliases: |
IEDA, PGR10 |
Location: |
Chr.3:154337672-154429715 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR149 | NM_001038705.1 | NP_001033794.1 | ||
| AY255534.1 | AAO85046.1 | |||
| DQ318360.1 | ABC49857.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763308 | Chr.3:154186065 - 154347347 on Build GRCh38 | Loss |
|
| nsv4073 | Chr.3:154313497 - 154345052 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map