Assay Details
Target Gene Details
Entrez Gene ID: | 57110 |
Gene Name: | HRAS like suppressor |
Gene Aliases: |
A-C1, H-REV107, HRASLS1, HRSL1, HSD28 |
Location: |
Chr.3:193241128-193281639 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HRASLS | NM_020386.4 | NP_065119.2 | ||
| XM_011513034.1 | XP_011511336.1 | |||
| XM_011513035.2 | XP_011511337.1 | |||
| XM_011513037.2 | XP_011511339.1 | |||
| XM_017006923.1 | XP_016862412.1 | |||
| AB030816.1 | BAB08110.1 | |||
| BC048095.1 | AAH48095.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003505 | Chr.3:193168634 - 193574329 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map