Assay Details
Target Gene Details
Entrez Gene ID: | 9117 |
Gene Name: | SEC22 homolog C, vesicle trafficking protein |
Gene Aliases: |
SEC22L3 |
Location: |
Chr.3:42547967-42601080 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC22C | NM_001201572.1 | NP_001188501.1 | ||
| NM_001201584.1 | NP_001188513.1 | |||
| NM_004206.3 | NP_004197.1 | |||
| NM_032970.3 | 7 | 5066 | NP_116752.1 | |
| XM_005265533.1 | 8 | 5240 | XP_005265590.1 | |
| XM_005265534.1 | 8 | 5140 | XP_005265591.1 | |
| XM_005265535.1 | 8 | 5198 | XP_005265592.1 | |
| XM_011534212.1 | 7 | 4918 | XP_011532514.1 | |
| XM_017007438.1 | XP_016862927.1 | |||
| XM_017007439.1 | XP_016862928.1 | |||
| AF039568.1 | AAD02171.1 | |||
| AK074576.1 | ||||
| AK293133.1 | ||||
| AY359041.1 | AAQ89400.1 | |||
| BC018437.1 | AAH18437.1 | |||
| BX377531.2 | ||||
| CR749670.1 | 5 | 4620 | CAH18461.1 |
More Information
Set Membership: |
|

Genomic Map