Assay Details
Target Gene Details
Entrez Gene ID: | 27333 |
Gene Name: | golgi integral membrane protein 4 |
Gene Aliases: |
GIMPC, GOLPH4, GPP130, P138 |
Location: |
Chr.3:168008672-168095925 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLIM4 | NM_001308155.1 | NP_001295084.1 | ||
| NM_014498.4 | NP_055313.1 | |||
| XM_005247364.2 | XP_005247421.1 | |||
| XM_005247365.2 | XP_005247422.1 | |||
| XM_017006183.1 | XP_016861672.1 | |||
| AK307557.1 | ||||
| U55853.1 | AAB58419.1 |
More Information
Set Membership: |
|

Genomic Map