Assay Details
Target Gene Details
Entrez Gene ID: | 440952 |
Gene Name: | EIF1B antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:40173145-40309698 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF1B-AS1 | NR_033965.1 | |||
| AK309874.1 |
Target Gene Details
Entrez Gene ID: | 25924 |
Gene Name: | myosin VIIA and Rab interacting protein |
Gene Aliases: |
SLAC2-C, SLAC2C |
Location: |
Chr.3:39808914-40260321 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYRIP | NM_001284423.1 | NP_001271352.1 | ||
| NM_001284424.1 | NP_001271353.1 | |||
| NM_001284425.1 | NP_001271354.1 | |||
| NM_001284426.1 | NP_001271355.1 | |||
| NM_015460.3 | NP_056275.2 | |||
| NR_104316.1 | ||||
| XM_011533575.1 | XP_011531877.1 | |||
| AB083783.1 | BAC15555.1 | |||
| AF396687.1 | AAM43954.1 | |||
| AK125334.1 | ||||
| AK126013.1 | ||||
| AK294714.1 | ||||
| AK312412.1 | ||||
| AK316068.1 | ||||
| BC092512.1 | AAH92512.1 | |||
| BC109311.1 | AAI09312.1 | |||
| BC109312.1 | AAI09313.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834667 | Chr.3:40149053 - 40317061 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map