Assay Details
Target Gene Details
Entrez Gene ID: | 2122 |
Gene Name: | MDS1 and EVI1 complex locus |
Gene Aliases: |
AML1-EVI-1, EVI1, MDS1, MDS1-EVI1, PRDM3, RUSAT2 |
Location: |
Chr.3:169083499-169663781 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
More Information
Set Membership: |
|

Genomic Map