Assay Details
Target Gene Details
Entrez Gene ID: | 26137 |
Gene Name: | zinc finger and BTB domain containing 20 |
Gene Aliases: |
DPZF, HOF, ODA-8S, PRIMS, ZNF288 |
Location: |
Chr.3:114314500-115147280 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB20 | NM_001164342.2 | NP_001157814.1 | ||
| NM_001164343.2 | NP_001157815.1 | |||
| NM_001164344.2 | NP_001157816.1 | |||
| NM_001164345.2 | NP_001157817.1 | |||
| NM_001164346.2 | NP_001157818.1 | |||
| NM_001164347.2 | NP_001157819.1 | |||
| NM_015642.5 | NP_056457.3 | |||
| NR_121662.1 | ||||
| AF139460.2 | AAG28340.2 | |||
| AK225788.1 | ||||
| AK290116.1 | ||||
| AK315307.1 | ||||
| AL050276.1 | CAB43377.1 | |||
| BC029041.1 | AAH29041.1 | |||
| BX647778.1 | CAH56190.1 | |||
| CB164748.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829679 | Chr.3:114331081 - 114533888 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map