Assay Details
Target Gene Details
Entrez Gene ID: | 131405 |
Gene Name: | tripartite motif containing 71 |
Gene Aliases: |
LIN-41, LIN41 |
Location: |
Chr.3:32818018-32897826 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TRIM71 | NM_001039111.2 | NP_001034200.1 | ||
DQ232881.1 | ABB18374.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv998302 | Chr.3:32595752 - 33112754 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1003285 | Chr.3:31386933 - 33172886 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
