Assay Details
Target Gene Details
Entrez Gene ID: | 57129 | 
Gene Name: | mitochondrial ribosomal protein L47 | 
Gene Aliases: | 
	
		CGI-204, L47mt, MRP-L47, NCM1 | 
Location: | 
		
			Chr.3:179588465-179604646 on Build GRCh38 | 
		Assay Gene Location: | Within Intron 6 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| MRPL47 | NM_020409.2 | NP_065142.2 | ||
| NM_177988.1 | NP_817125.1 | |||
| AY212270.1 | AAO92749.1 | |||
| BC021575.1 | ||||
| BC032522.1 | AAH32522.1 | |||
| CA428787.1 | 
More Information
Set Membership: | 
		   
					
						
							 | 
				

Genomic Map