Assay Details
Target Gene Details
Entrez Gene ID: | 58477 |
Gene Name: | SRP receptor beta subunit |
Gene Aliases: |
APMCF1 |
Location: |
Chr.3:133784033-133821492 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRPRB | NM_021203.3 | NP_067026.3 | ||
| AF086914.1 | AAP97144.1 | |||
| AF141882.3 | AAD34888.3 | |||
| AK027307.1 | ||||
| AK027525.1 | BAB55176.1 | |||
| AK075531.1 | BAC11675.1 | |||
| BC063001.1 | AAH63001.1 | |||
| BC065299.1 | AAH65299.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829729 | Chr.3:133726027 - 133901543 on Build GRCh38 | Loss |
|
| nsv4018 | Chr.3:133767023 - 133811824 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map