Assay Details
Target Gene Details
Entrez Gene ID: | 91775 |
Gene Name: | neurexophilin and PC-esterase domain family member 3 |
Gene Aliases: |
FAM55C, MST115, MSTP115 |
Location: |
Chr.3:101779185-101828231 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NXPE3 | NM_001134456.1 | NP_001127928.1 | ||
| NM_145037.2 | NP_659474.1 | |||
| AK054664.1 | BAB70787.1 | |||
| AK289689.1 | ||||
| AY358207.1 | AAQ88574.1 | |||
| BC009431.1 | AAH09431.1 |

Genomic Map