Assay Details
Target Gene Details
Entrez Gene ID: | 8927 |
Gene Name: | bassoon presynaptic cytomatrix protein |
Gene Aliases: |
ZNF231 |
Location: |
Chr.3:49554489-49671549 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BSN | NM_003458.3 | NP_003449.2 | ||
| AF052224.1 | AAC83555.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508217 | Chr.3:49578013 - 49732463 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map