Assay Details
Target Gene Details
Entrez Gene ID: | 1371 |
Gene Name: | coproporphyrinogen oxidase |
Gene Aliases: |
CPO, CPX, HCP |
Location: |
Chr.3:98568448-98593684 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPOX |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834775 | Chr.3:98451215 - 98648708 on Build GRCh38 | Gain |
|
| nsv591065 | Chr.3:98485354 - 99047482 on Build GRCh38 | Gain |
|
| dgv293n21 | Chr.3:98382257 - 98598992 on Build GRCh38 | Loss |
|
| dgv4819n100 | Chr.3:98565439 - 98662629 on Build GRCh38 | Gain |
|

Genomic Map