Assay Details
Target Gene Details
Entrez Gene ID: | 407738 |
Gene Name: | family with sequence similarity 19 member A1, C-C motif chemokine like |
Gene Aliases: |
TAFA-1, TAFA1 |
Location: |
Chr.3:67991592-68546056 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM19A1 | NM_001252216.1 | NP_001239145.1 | ||
| NM_213609.3 | NP_998774.2 | |||
| XM_017006386.1 | XP_016861875.1 | |||
| XM_017006387.1 | XP_016861876.1 | |||
| XM_017006388.1 | XP_016861877.1 | |||
| XM_017006389.1 | XP_016861878.1 | |||
| XM_017006390.1 | XP_016861879.1 | |||
| AK289668.1 | ||||
| AK290446.1 | ||||
| AL713702.1 | CAD28501.1 | |||
| AY325114.1 | AAP92406.1 | |||
| BC025746.1 | AAH25746.1 | |||
| DA485098.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003158 | Chr.3:66999779 - 68395539 on Build GRCh38 | Loss |
|
| esv999186 | Chr.3:68017838 - 68032369 on Build GRCh38 | Insertion |
|
| esv3596476 | Chr.3:68027068 - 68027902 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map