Assay Details
Target Gene Details
Entrez Gene ID: | 104797538 |
Gene Name: | KCNMB2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:178525467-178860405 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNMB2-AS1 | NR_126560.1 | 8 | 1940 | |
| NR_126561.1 | 6 | 1681 | ||
| AF279780.1 | 6 | 1679 | ||
| AY679731.1 | 1 | 725 | ||
| AY679732.1 | 1 | 433 | ||
| U61096.1 | 1 | 98 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3569136 | Chr.3:178522284 - 178539169 on Build GRCh38 | Loss |
|

Genomic Map