Assay Details
Target Gene Details
Entrez Gene ID: | 23007 |
Gene Name: | phospholipase C eta 1 |
Gene Aliases: |
PLCL3 |
Location: |
Chr.3:155458603-155745723 on Build GRCh38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLCH1 | XM_017005927.1 | 24 | 5789 | XP_016861416.1 |
| AK022610.1 | BAB14129.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3598273 | Chr.3:155342999 - 155574230 on Build GRCh38 | Loss |
|
| esv3598274 | Chr.3:155391781 - 155464589 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map